L(+)-Glutamine cell culture reagent
Catalog # AAJ60573-A1
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:1 kg
- Environmentally Preferable:
- Cat. No.:AAJ60573-A1
- Supplier no.:J60573-A1
Specifications
About this item
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
:
:146.15 g/mol
Boiling Pt:355
Melting Pt:185…186 °C
Storage Temperature:Ambient
EINECS:200-292-1
CAS Number:56-85-9
Formula:C₅H₁₀N₂O₃
MDL Number:MFCD00008044
Merck Index:14,4471