Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific

Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific
 
Phosphate, 0.2 M buffer soln., pH 4.4, Thermo Scientific

 

 

Liquid

CAS No.: 7558-80-7
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00003527

 

 

 

  • Catalog No:
  • AAJ60467-AP
  • AAJ60467-K2
  • Physical Form:
  • Liquid
  • Liquid
  • CAS No.:
  • 7558-80-7
  • 7558-80-7
  • MDL No.:
  • MFCD00003527
  • MFCD00003527
  • Storage Temperature:
  • 2°C to 8°C
  • 2°C to 8°C

 

Product Family Options

 

  • Size
     
     
  • 500 mL
       
       
    Each (500ml)
    Each (500ml) - $101.92
    $101.92
    Each (500ml)
       $101.92
     

     : J60467-AP
     
     
    Specifications
    Cat. No.
    AAJ60467-AP
    Physical Form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527
    Storage Temperature
    2°C to 8°C
  • 1 L
       
       
    Each (1l)
    Each (1l) - $150.78
    $150.78
    Each (1l)
       $150.78
     

     : J60467-K2
     
     
    Specifications
    Cat. No.
    AAJ60467-K2
    Physical Form
    Liquid
    CAS No.
    7558-80-7
    MDL No.
    MFCD00003527
    Storage Temperature
    2°C to 8°C