Phosphate, 0.2 M buffer soln., pH 6.8, Thermo Scientific
AAJ60325-K2
:
- CAS No.:7558-80-7
- Size:1 L
- MDL No.:MFCD00003527
- Physical Form:Liquid
- Cat. No.:AAJ60325-K2
- Supplier no.:J60325-K2
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527