$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Buffers
Assay Buffers
Kinase Buffer II (5X), Thermo Scientific
Kinase Buffer II (5X), Thermo Scientific
:
Thermo Scientific Chemicals
Kinase Buffer II (5X), Thermo Scientific
:
Thermo Scientific Chemicals
Liquid
Physical Form:
Liquid
Alfa Aesar_Buy 3 Get 2 Complimentary items
msds_ALFAJ60269