Sodium borate, 0.5 M buffer soln., pH 8.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 1330-43-4
Physical Form: Liquid
MDL No.: MFCD00081185
Specifications
- CAS No.:1330-43-4
- MDL No.:MFCD00081185
- Physical Form:Liquid