Sodium bicarbonate, 1 M buffer soln., pH 9.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Liquid
CAS No.: 144-55-8
Physical Form: Liquid
MDL No.: MFCD00003528
Specifications
- CAS No.:144-55-8
- MDL No.:MFCD00003528
- Physical Form:Liquid