Methyl-4-fluoro-2-methoxybenzoate 97+%
About this item
Purity: 97+% CAS No.: 204707-42-6 Molecular Formula: C9H9FO3 Formula Weight: 184.17 MDL No.: MFCD06203990
- CAS Number:204707-42-6
- Formula:C₉H₉FO₃
- MDL Number:MFCD06203990
- Molecular Weight:184.17
- Storage Temperature:Ambient
1 Options Available Below
Product Details & Documents
Purity: 97+% CAS No.: 204707-42-6 Molecular Formula: C9H9FO3 Formula Weight: 184.17 MDL No.: MFCD06203990
- CAS Number:204707-42-6
- Formula:C₉H₉FO₃
- MDL Number:MFCD06203990
- Molecular Weight:184.17
- Storage Temperature:Ambient
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- 250mgDiscontinued-Supplier Number: -
- 1gDiscontinued-Supplier Number: -
- Supplier Number: H52370-06
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



