Order Entry
Puerto Rico
ContactUsLinkComponent
Agarose 6% B - CL, cross-linked, beads
Flammable
Agarose 6% B - CL, cross-linked, beads
Catalog # AAH32568-AU
CAS Number:  
Flammable
Agarose 6% B - CL, cross-linked, beads
Catalog # AAH32568-AU
Supplier Number:  H32568-AU
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1000 ml
  • Cat. No.:
    AAH32568-AU
  • Supplier no.:
    H32568-AU

Specifications

About this item

 :
Boiling Pt:100 °C
Density:1.040
Flash Pt:38 °C(100 °F)
Flash Pt:38 °C (100 °F)
Storage Temperature:Ambient
UN:1170
ADR:"3",III
 :GHS02
CAS Number:9012-36-6
MDL Number:MFCD00081294
ADR:3,III