


Sulfuryl chloride 1 M in dichloromethane
Catalog # AAH31899-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:AAH31899-AP
- Supplier no.:H31899-AP
Specifications
About this item
:
:134.97 g/mol
Density:1.352
Storage Temperature:Ambient
EINECS:232-245-6
UN:2922
ADR:"8",III
:GHS05
:GHS06
:GHS08
CAS Number:7791-25-5
Formula:Cl₂O₂S
MDL Number:MFCD00011451
ADR:8,III