Order Entry
Puerto Rico
ContactUsLinkComponent
Zinc, Quantofix® Test Strips, Thermo Scientific
Zinc, Quantofix® Test Strips, Thermo Scientific
Catalog # AA46651-QA
CAS Number:  
Zinc, Quantofix® Test Strips, Thermo Scientific
Catalog # AA46651-QA
Supplier Number:  046651-QA
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Zinc Quantofix® Test Strips
  • Size:
    1 kit
  • UN No.:
    1824
  • Cat. No.:
    AA46651-QA
  • Supplier no.:
    46651-QA
  • No. of tests:
    100

Specifications

About this item

Zinc, Quantofix® Test Strips are used to measure zinc in solutions quickly

UN No.:UN1824

It is used to regularly check the plating baths in the metal plating industry. It is also used to check the concentration of zinc in cooling water treatment, where zinc salts are used as corrosion inhibitors.