To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Size:100 ml
- MDL No.:MFCD03792350
- Cat. No.:AAA44789-AE
- Supplier no.:44789-AE
These standards are applicable for the determination of elements as dissolved elements in ground waters, surface waters, and drinking water, and total recoverable elements in these waters as well as wastewaters, sludges, and soils samples.
Matrix: 5% HNO3
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD03792350
Concentration: Bi 10 µg/mL ±2%; In 10 µg/mL ±2%; Lu 10 µg/mL ±2%; Sc 10 µg/mL ±2%; Tb 10 µg/mL ±2%; Y 10 µg/mL ±2%; ₆Li 10 µg/mL ±2%