Order Entry
Puerto Rico
ContactUsLinkComponent
Wall-Mountable Large Apparel Dispenser, PVC/Clear Acrylic, TrippNT
Wall-Mountable Large Apparel Dispenser, PVC/Clear Acrylic, TrippNT
Catalog # 10012-628
Supplier:  TrippNT
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Wall-Mountable Large Apparel Dispenser, PVC/Clear Acrylic, TrippNT
Catalog # 10012-628
Supplier:  TrippNT
Supplier Number:  50101
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Specifications

  • Dispenser type:
    Wall-mountable
  • Material:
    Acrylic
  • No. of Compartments:
    2
  • PPE type:
    Multi-use
  • Description:
    Wall-mountable dispenser with two large compartments
  • Weight:
    5 kg (11 lbs.)
  • Cat. No.:
    10012-628
  • Supplier no.:
    50101

Specifications

About this item

2-in-1 white PVC/clear acrylic wall-mountable large apparel dispenser with two large compartments dispenses from bottom and loads from the top

It can be used for anything from hairnets, safety apparel and shoe covers to wet wipes and goggles. Mounts easily with keyholes wherever you need it most. Provides easy access to personal protection supplies, disposables, or consumables.