About this item
Used in Western blotting protocols.
- Contaminant Testing:
- DNase (endo): Passes test
- DNase (exo): Passes test
- Protease: Passes test
20X TBS (Tris Buffered Saline) solution consisting of 500mM Tris, 60 mM KCl, 2.8 M NaCl, and 1.0% Tween 20 in high purity dH₂O adjusted to pH 7.4.
Solution is 0.2 μm filtered and is supplied in an upright carton with a convenient dispensing spout.
Dilute to 1X with high purity water for a solution consisting of 25 mM Tris, pH 7.4, 3.0 mM KCl, 140 mM NaCl and 0.05% Tween 20.
Storage temperature of solution: Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Storage Temperature:Ambient