ContactUsLinkComponent
Dilatometer, Volumetric, Ace Glass Incorporated
  89054-078
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Dilatometer, Volumetric, Ace Glass Incorporated
  89054-078
 :  Ace Glass
 :  6284-10
Product available on GSA Advantage®
Small Business Enterprise
  • Description:
    VolumeVolumetric Dilatometertric Dilatometer
  • Cat. No.:
    89054-078
  • Supplier no.:
    6284-10
This dilatometer is designed for use in any reaction where there is a change in volume
Described in Experiments in Physical Chemistry, Shoemaker & Garland, 1962, by McGraw-Hill Book Company, Inc. Experiment 27. Capacity of bath is 75mL, bulb 50mL, with capillary tube graduated from 0.0cm at bottom to 15.0cm at top. The capillary I.D. is 0.6mm. Stopcock is 2mm bore. Fabricated from borosilicate glass.