Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VEGFC Goat Polyclonal Antibody
Anti-VEGFC Goat Polyclonal Antibody
Catalog # 89363-726
Supplier:  Genetex
Anti-VEGFC Goat Polyclonal Antibody
Catalog # 89363-726
Supplier:  Genetex
Supplier Number:  GTX59592

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Vascular Endothelial Growth Factor C
  • Antigen Symbol:
    VEGFC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 μg
  • Gene ID:
    7424
  • Antigen Synonyms:
    Vascular Endothelial Growth Factor C|VEGF C|VRP|Vascular endothelial growth factor-related protein C
  • Storage Buffer:
    phosphate buffered saline (PBS), pH 7.2, containing 30% glycerol, 0.5% BSA, 0.01% thimerosal.
  • Storage Temperature:
    The antibody solution should be gently mixed before use. Store at +4C. Product should remain stable for at least 12 months.
  • Concentration:
    0.5 mg/ml
  • Tested Applications:
    IHC, WB
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    89363-726
  • Supplier no.:
    GTX59592

Specifications

About this item

Goat polyclonal antibody to VEGF-C

Western blotting (0.5-1 µg/ml) and Immunohistochemistry with both frozen and paraffin sections (15-25 µg/ml). However, the optimal concentrations should be determined individually. The antibody recognizes human VEGF-C. Reactivity to other species has not been tested.

Type: Primary
Antigen: VEGFC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human