Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-TNFSF15 Rabbit Polyclonal Antibody
Anti-TNFSF15 Rabbit Polyclonal Antibody
  89355-530
 :  Genetex
Anti-TNFSF15 Rabbit Polyclonal Antibody
  89355-530
 :  Genetex
 :  GTX108463

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Tumor Necrosis Factor Superfamily Member 15
  • Antigen Symbol:
    TNFSF15
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    9966
  • Antigen Synonyms:
    TL1A|Vascular Endothelial Cell Growth Inhibitor|VEGI192A|VEGI|TNLG1B|TNF Ligand-Related Molecule 1
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    28 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TL1A. The exact sequence is proprietary.
  • Tested Applications:
    WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89355-530
  • Supplier no.:
    GTX108463

 

 

Rabbit polyclonal antibody to TL1A (tumor necrosis factor (ligand) superfamily, member 15)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TNFSF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human