Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-THOC7 Rabbit Polyclonal Antibody
Anti-THOC7 Rabbit Polyclonal Antibody
Catalog # 89359-082
Supplier:  Genetex
Anti-THOC7 Rabbit Polyclonal Antibody
Catalog # 89359-082
Supplier:  Genetex
Supplier Number:  GTX119877

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    THO Complex 7
  • Antigen Symbol:
    THOC7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    80145
  • Antigen Synonyms:
    hTREX30|NIF3L1BP1|fSAP24
  • Storage Buffer:
    1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    24 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human THOC7. The exact sequence is proprietary.
  • Tested Applications:
    ICC/IF, WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89359-082
  • Supplier no.:
    GTX119877

Specifications

About this item

Component of the THO subcomplex of the TREX complex. The TREX complex specifically associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism. Binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5' end of the mRNA where it functions in mRNA export. The recruitment occurs via an interaction between THOC4 and the cap-binding protein NCBP1. UAP56 functions as a bridge between THOC4 and the THO complex. The TREX complex is essential for the export of Kaposi's sarcoma-associated herpesvirus (KSHV) intronless mRNAs and infectious virus production. The recruitment of the TREX complex to the intronless viral mRNA occurs via an interaction between KSHV ORF57 protein and THOC4.

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: THOC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human