To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumor-associated glycoprotein 72
- Antigen Symbol:TAG72
- Clonality:Monoclonal
- Clone:B72.3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:500 µL
- Storage Buffer:Tissue culture supernatant containing sodium azide
- Storage Temperature:+4°C
- Immunogen:Membrane-enriched fraction of a human breast carcinoma liver metastasis
- Tested Applications:IHC-P
- Purification:unpurified
- Cat. No.:89351-528
- Supplier no.:GTX20691
Specifications
About this item
The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for B72.3 staining. About 60% of carcinoma patients express TAG-72 in their sera.
Type: Primary
Antigen: TAG72
Clonality: Monoclonal
Clone: B72.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human