Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Snta1 Rabbit Polyclonal Antibody
Anti-Snta1 Rabbit Polyclonal Antibody
Catalog # 89356-876
Supplier:  Genetex
Anti-Snta1 Rabbit Polyclonal Antibody
Catalog # 89356-876
Supplier:  Genetex
Supplier Number:  GTX11187

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Syntrophin Alpha 1
  • Antigen Symbol:
    SNTA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    20648
  • Antigen Synonyms:
    TACIP1|SNT1|LQT12|dJ1187J4.5
  • Storage Buffer:
    Preservative: 15mM Sodium Azide; Constituents: 0.01M PBS. pH 7.4
  • Storage Temperature:
    For long term storage, please keep at –20 °C. Avoid repeared freeze-thaw cycles.
  • Immunogen:
    Synthetic peptide conjugated to KLH, corresponding to amino acids 191-206 of Mouse Syntrophin alpha 1.
  • Tested Applications:
    IHC, WB
  • Purification:
    IgG fraction
  • Cat. No.:
    89356-876
  • Supplier no.:
    GTX11187

Specifications

About this item

Rabbit polyclonal to Syntrophin alpha 1

IHC: Use at a dilution of 1/1000 (determined by immunohistochemistry using paraformaldehyde-fixed, Triton X-100 treated, frozen sections of rat skeletal muscle). WB: Use at a dilution of 1/2000 (determined by immunoblotting using a membrane fraction extract of rat skeletal muscle homogenate). Predicted molecular weight: 60 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Snta1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit