Specifications
- Antibody Type:Primary
- Antigen Name:Syntaxin 2
- Antigen Symbol:STX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Drosophila
- Western Blot:Yes
- Size:50 μg
- Gene ID:25130
- Antigen Synonyms:STX2B|EPIM|STX2C|EPM|STX2A
- Storage Buffer:Phosphate-buffered saline, pH 7.2, containing 0.09% sodium azide and 50% glycerol
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide: MRDRLPDLTACRKSDDGDN conjugated to KLH, corresponding to amino acids 1-19 of Rat Syntaxin 2.
- Tested Applications:IP, WB
- Purification:Immunogen affinity purified
- Cat. No.:89359-354
- Supplier no.:GTX12369
Specifications
About this item
Syntaxin 2, also known as epimorphin, is a 35 kDa type II integral membrane that belongs to the t SNARE family, a group of proteins involved in protein transport. Syntaxin 2 was first identified as a factor that promotes branching morphogenesis in mammary epithelial cells. Studies using pancreatic exocrine acinar cells, in which the exocytic pathways are well defined, show primary localization of syntaxin 2 to the apical plasma membrane. Homologs of syntaxin 2 have been identified in various eukaryotes ranging from yeast to human. Neuronal syntaxin is a component of a 20S complex implicated in vesicle docking and fusion, which involves its interaction with syntaxin through a ATP dependent reaction modulated by synaptotagmin, SNAP 25, alpha SNAP, and NSF.
For IP: Use at a concentration of 10 ?g/ml. For WB: Use at a concentration of 1 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: STX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila