Anti-PRPF4 Rabbit Polyclonal Antibody
89359-146
: Genetex
- Antibody Type:Primary
- Antigen Name:Pre-mRNA Processing Factor 4
- Antigen Symbol:PRPF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:9128
- Antigen Synonyms:SNRNP60|PRP4|HPRP4P|HPRP4|Prp4p|RP70
- Storage Buffer:0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:59 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human PRPF4. The exact sequence is proprietary.
- Tested Applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89359-146
- Supplier no.:GTX120537
The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.[supplied by OMIM]
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: PRPF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human