Anti-LTB4R Rabbit Polyclonal Antibody
89355-330
: Genetex
- Antibody Type:Primary
- Antigen Name:Leukotriene B4 Receptor
- Antigen Symbol:LTB4R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:1241
- Antigen Synonyms:LTBR1|LTB4R1|GPR16|CMKRL1|P2Y7|P2RY7|BLT1|BLTR
- Storage Buffer:0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:38 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human P2Y7. The exact sequence is proprietary.
- Tested Applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89355-330
- Supplier no.:GTX108169
Rabbit polyclonal antibody to P2Y7 (leukotriene B4 receptor)
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: LTB4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human