Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NMDA Receptor 3A + 3B Mouse Monoclonal Antibody [clone: 0]
Anti-NMDA Receptor 3A + 3B Mouse Monoclonal Antibody [clone: 0]
  89361-314
 :  Genetex
Anti-NMDA Receptor 3A + 3B Mouse Monoclonal Antibody [clone: 0]
  89361-314
 :  Genetex
 :  GTX22639

 

  • Antibody Type:
    Primary
  • Antigen Name:
    NMDA Receptor 3A + 3B
  • Antigen Symbol:
    NMDA Receptor 3A + 3B
  • Clonality:
    Monoclonal
  • Clone:
    0
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Amino Acid Number:
    780 to 914
  • Storage Buffer:
    Preservative: None; Constituents: PBS. pH 7.4
  • Storage Temperature:
    -20 °C to -70 °C
  • Immunogen:
    Fusion protein, corresponding to amino acids 780-914 from the intracellular IIIIV loop of Human NR3A + NR3B.
  • Tested Applications:
    WB
  • Purification:
    purified by Protein G chromatography
  • Cat. No.:
    89361-314
  • Supplier no.:
    GTX22639

 

 

Two new subunits of the NMDA subtype of glutamate receptor (NMDAR) have been identified, NR3A and NR3B, with NR3B likely being the final member of the NMDAR family. These subunits show high sequence homology, although different distribution patterns: NR3B is expressed predominately in motor neurons, while NR3A is more widely distributed. When expressed in Xenopus oocytes, NR3A or NR3B assemble with NR1 to form excitatory glycine receptors that are unaffected by glutatmate.

Type: Primary
Antigen: NMDA Receptor 3A + 3B
Clonality: Monoclonal
Clone: 0
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human