Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Ngf Goat Polyclonal Antibody
Anti-Ngf Goat Polyclonal Antibody
Catalog # 89355-482
Supplier:  Genetex
Anti-Ngf Goat Polyclonal Antibody
Catalog # 89355-482
Supplier:  Genetex
Supplier Number:  GTX10837

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Nerve Growth Factor
  • Antigen Symbol:
    NGF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    25μg
  • Neutralisation:
    Yes
  • Gene ID:
    310738
  • Antigen Synonyms:
    HSAN5|Beta-NGF|NGFB
  • Storage Buffer:
    PBS, pH7.4 containing 5% trehalose
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.1mg/ml
  • Immunogen:
    Recombinant full length rat NGF beta protein expressed in Sf 21 insect cells.
  • Tested Applications:
    ELISA, Neut, WB
  • Purification:
    Immunogen affinity purified
  • Cat. No.:
    89355-482
  • Supplier no.:
    GTX10837

Specifications

About this item

Goat polyclonal to NGF beta

ELISA: Use at a concentration of 0.5-1 ?g/ml. Neut: Use at a concentration of 0.05-0.15 ?g/ml. WB: Use at a concentration of 0.1-0.2 ?g/ml. Detects a band of approximately 13.2 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Ngf
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat