Anti-NTF4 Mouse Monoclonal Antibody [clone: 0]
89352-604
: Genetex
- Antibody Type:Primary
- Antigen Name:Neurotrophin 4
- Antigen Symbol:NTF4
- Clonality:Monoclonal
- Clone:0
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:500 μg
- Gene ID:4909
- Antigen Synonyms:NTF5|neurotrophin 5|NT-4/5|neurotrophic factor 5|neurotrophin 5 (neurotrophin 4/5) |neurotrophin 4|NT5|neurotrophic factor 4|NTF4|NT4
- Storage Buffer:Phosphate buffered saline
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Concentration:1 mg/ml
- Immunogen:Recombinant full length protein (Human).
- Tested Applications:ELISA
- Purification:Protein A purified
- Cat. No.:89352-604
- Supplier no.:GTX29330
Mouse monoclonal to Neurotrophin 4
ELISA: Use at a dilution of 1/10000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NTF4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human