Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]
  89358-754
 :  Genetex
Anti-MPO Mouse Monoclonal Antibody [clone: 2C7]
  89358-754
 :  Genetex
 :  GTX11729

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Myeloperoxidase
  • Antigen Symbol:
    MPO
  • Clonality:
    Monoclonal
  • Clone:
    2C7
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    50 μg
  • Gene ID:
    4353
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin
  • Storage Temperature:
    Store at +4 °C or at –20 °C. This product should be stored undiluted. Storage in frost free freezers is not recommended. This product is photosensitive. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    Human myeloperoxidase.
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G
  • Cat. No.:
    89358-754
  • Supplier no.:
    GTX11729

 

 

Myeloperoxidase is a hemoprotein that is abundantly expressed in neutrophils and secreted during their activation. Native Myeloperoxidase is represented as a covalently bound tetrameric complex of two glycosylated alpha chains (MW 59 - 64 kDa) and two unglycosylated beta chains (MW 14 kDa) with total MW 150 kDa and theoretical pI 9.2. Traditionally Myeloperoxidase was considered as a main target of anti-neutrophil cytoplasm antibodies (ANCA), the serological markers for certain systemic vasculities e.g. periarteriitis nodosa, microscopic polyarteriitis and pulmonary eosinophilic granulomatosis (Churg-Strauss syndrome). Low to moderate anti-Myeloperoxidase autoantibody levels are also reported in rheumatoid arthritis. Recently it was shown that Myeloperoxidase participates in the initiation and progression of cardiovascular disease. It possesses potent proinflammatory properties and may contribute directly to tissue injury. Now Myeloperoxidase is under consideration as one of the most promising cardiac markers.

Recommended Starting Dilutions:Flow Cytometry: Neat - 1/10Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human