Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-HLA-G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-G/9]
Anti-HLA-G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-G/9]
  89353-838
 :  Genetex
Anti-HLA-G Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-G/9]
  89353-838
 :  Genetex
 :  GTX78335

 

  • Antibody Type:
    Primary
  • Antigen Name:
    HLA class I histocompatibility antigen, alpha chain G
  • Antigen Symbol:
    HLA-G
  • Clonality:
    Monoclonal
  • Clone:
    MEM-G/9
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 μg
  • Gene ID:
    3135
  • Antigen Synonyms:
    HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
  • Storage Buffer:
    The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
  • Storage Temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
  • Concentration:
    0.1 mg/ml
  • Immunogen:
    Recombinant human HLA-G refolded with beta2-microglobulin and peptide.
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Cat. No.:
    89353-838
  • Supplier no.:
    GTX78335

 

 

Monoclonal Antibody to HLA-G (Human) Phycoerythrin (PE)

The reagent is designed for Flow Cytometry analysis of cells expressing HLA-G molecule on the cell surface. Suggested working dilution is 1:100. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: HLA-G
Clonality: Monoclonal
Clone: MEM-G/9
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human