Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Anti-E2F1 Rabbit Polyclonal Antibody

Anti-E2F1 Rabbit Polyclonal Antibody

50 μg
Catalog # 89367-394
Supplier:   Genetex
Supplier #:  GTX14770
Anti-E2F1 Rabbit Polyclonal Antibody
Catalog # 89367-394
Supplier:  Genetex
Anti-E2F1 Rabbit Polyclonal Antibody

50 μg

Price
$634.19
Log in for availability
Each (50µG)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

E2F's are DNA binding proteins, which associate with negative regulators, such as the retinoblastoma p107 protein, resulting in an altered rate of gene transcription. E2F1 is proposed to be involved in several cellular processes that range from tumor suppressor, cell progression and oncogenesis. E2F1 overexpression can also drive cells into apoptosis.

IHC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000. Detects a band of approximately 53 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: E2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications
  • Size:
    50 μg
  • Clonality:
    Polyclonal
  • ImmunoPrecipitation:
    Yes
  • Purification:
    Immunogen affinity purified
  • Storage Buffer:
    Preservative: 0.02% Thimerosal / MerthiolateConstituents: PBS
  • Tested Applications:
    IHC, IP, WB
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Concentration:
    0.5 mg/ml
  • Cat. No.:
    89367-394
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Antibody Type:
    Primary
  • Antigen Symbol:
    E2F1
  • Gene ID:
    1869
  • Immunogen:
    Synthetic peptide: SGLLPEEFISLSPPHEALDYHFG, corresponding to amino acids 392-414 of Human E2F1.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen Synonyms:
    Retinoblastoma binding protein 3|RBBP-3|PRB-binding protein E2F-1|Retinoblastoma-associated protein 1|RBAP-1|E2F-1|Transcription factor E2F1|E2F1|PBR3|RBBP3
  • Antigen Name:
    E2F Transcription Factor 1
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.