Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CYBRD1 Rabbit Polyclonal Antibody
Anti-CYBRD1 Rabbit Polyclonal Antibody
Catalog # 89350-554
Supplier:  Genetex
undefined
Anti-CYBRD1 Rabbit Polyclonal Antibody
Catalog # 89350-554
Supplier:  Genetex
Supplier Number:  GTX104793

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Cytochrome b Reductase 1
  • Antigen Symbol:
    CYBR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    79901
  • Antigen Synonyms:
    CYB561A2|Ferric-chelate reductase 3|CYBRD1|Cytochrome b reductase 1|DCYTB|FRRS3|Duodenal cytochrome b
  • Storage Buffer:
    1XPBS, 40% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    32 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at -20oC or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Cytochrome b reductase 1. The exact sequence is proprietary.
  • Tested Applications:
    WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89350-554
  • Supplier no.:
    GTX104793

Specifications

About this item

Rabbit polyclonal antibody to Cytochrome b reductase 1 (cytochrome b reductase 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CYBRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: