Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-TPP1 Rabbit Polyclonal Antibody
  89350-140
 :  Genetex
undefined
Anti-TPP1 Rabbit Polyclonal Antibody
  89350-140
 :  Genetex
 :  GTX103896

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Tripeptidyl-peptidase 1
  • Antigen Symbol:
    TPP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    1200
  • Antigen Synonyms:
    TPP-1|CLN2|GIG1|SCAR7|LPIC
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    61 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    1mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human TPP1. The exact sequence is proprietary.
  • Tested Applications:
    WB
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89350-140
  • Supplier no.:
    GTX103896

 

 

Rabbit Polyclonal antibody to TPP1 (tripeptidyl peptidase I)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TPP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human