Anti-CG beta 2 Mouse Monoclonal Antibody [clone: INN-hCG-22]
89367-954
: Genetex
- Antibody Type:Primary
- Antigen Name:Chorionic Gonadotropin, Beta Polypeptide
- Antigen Symbol:CG beta 2
- Clonality:Monoclonal
- Clone:INN-hCG-22
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:1082
- Antigen Synonyms:CGB3|CGB8|CGB5|hCGB|CGB7
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:hCG
- Tested Applications:ELISA, IHC-P, WB
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Cat. No.:89367-954
- Supplier no.:GTX76075
Mouse monoclonal antibody [INN-hCG-22] to CG beta
Recommended Starting Dilutions: ELISA: 1/100 - 1/500 Western Blotting: Max Dilution Non-reducing conditions Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CG beta 2
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: