To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Erb-B2 Receptor Tyrosine Kinase 2
- Antigen Symbol:C-ERBB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:6ml
- Environmentally Preferable:
- Gene ID:2064
- Antigen Synonyms:NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:Keyhole limpet hemocyanin conjugated synthetic human peptide c-erbB-2 oncoprotein.
- Tested Applications:AF, IHC-F
- Cat. No.:89351-294
- Supplier no.:GTX16932
This gene encodes a member of the epidermal growth factor (EGF) receptor family of receptor tyrosine kinases. This protein has no ligand binding domain of its own and therefore cannot bind growth factors. However, it does bind tightly to other ligand-bound EGF receptor family members to form a heterodimer, stabilizing ligand binding and enhancing kinase-mediated activation of downstream signalling pathways, such as those involving mitogen-activated protein kinase and phosphatidylinositol-3 kinase. Allelic variations at amino acid positions 654 and 655 of isoform a (positions 624 and 625 of isoform b) have been reported, with the most common allele, Ile654/Ile655, shown here. Amplification and/or overexpression of this gene has been reported in numerous cancers, including breast and ovarian tumors. Alternative splicing results in several additional transcript variants, some encoding different isoforms and others that have not been fully characterized.
AF: Use at an assay dependent dilution. IHC-F: Use at an assay dependent dilution. Suggested incubation period of 30 minutes at room temperature. Depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Although not required, heat treatment with 10 mM Citrate buffer, ph 6.0 may enhance the immunostaining in the overfixed tissues. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: C-ERBB2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: