Anti-CD79B Mouse Monoclonal Antibody [clone: AT105-1]
Catalog # 89353-390
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Cluster of Differentiation 79a molecule, immunoglobulin-associated beta
- Antigen Symbol:CD79B
- Clonality:Monoclonal
- Clone:AT105-1
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:1 ml
- Gene ID:974
- Antigen Synonyms:immunoglobulin-associated B29 protein|B29|RP23-418O11.7|Ig-beta|Igbeta|immunoglobulin-associated beta|B-cell-specific glycoprotein B29|B-cell antigen receptor complex-associated protein beta chain|Igb
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
- Storage Temperature:Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Peptide corresponding to an extracellular region of CD79 beta
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Purified IgG prepared by affinity chromatography on Protein A from tissue culture supernatant
- Cat. No.:89353-390
- Supplier no.:GTX75967
Specifications
About this item
Mouse monoclonal antibody [AT105-1] to CD79b
Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: AT105-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: