Anti-CD8 Mouse Monoclonal Antibody [clone: MRC OX-8]
89353-396
: Genetex
- Antibody Type:Primary
- Antigen Name:Cluster of differentiation 8
- Antigen Symbol:CD8
- Clonality:Monoclonal
- Clone:MRC OX-8
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:50 μg
- Gene ID:24930
- Antigen Synonyms:CD8alpha|p32|Leu-2|CD8a|MAL|CD8|CD8b|CD8beta
- Storage Buffer:PBS, pH7.2 containing BSA and 0.02% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Concentration:0.1 mg/ml
- Immunogen:High molecular weight rat thymocyte glycoproteins
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Protein G affinity purified
- Cat. No.:89353-396
- Supplier no.:GTX76219
Mouse monoclonal antibody [MRC OX-8] to CD8 alpha
For FACS: Use 1 ?g to label 10^6 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: