Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-NTPDase 1 Mouse Monoclonal Antibody [clone: A1]
Anti-NTPDase 1 Mouse Monoclonal Antibody [clone: A1]
Catalog # 89353-328
Supplier:  Genetex
Anti-NTPDase 1 Mouse Monoclonal Antibody [clone: A1]
Catalog # 89353-328
Supplier:  Genetex
Supplier Number:  GTX74823

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Ectonucleoside Triphosphate Diphosphohydrolase 1
  • Antigen Symbol:
    NTPDase 1
  • Clonality:
    Monoclonal
  • Clone:
    A1
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    100 test
  • Gene ID:
    953
  • Antigen Synonyms:
    Ectoapyrase|CD39|Ecto-apyrase|ENTPD1|SPG64
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
  • Storage Temperature:
    Store at 4°C. This product should be stored undiluted. DO NOT FREEZE. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    PHA activated human lymphocytes
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. No.:
    89353-328
  • Supplier no.:
    GTX74823

Specifications

About this item

Mouse monoclonal antibody [A1] to CD39

Type: Primary
Antigen: NTPDase 1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0