Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD274 Rabbit Polyclonal Antibody
Anti-CD274 Rabbit Polyclonal Antibody
Catalog # 89350-532
Supplier:  Genetex
undefined
Anti-CD274 Rabbit Polyclonal Antibody
Catalog # 89350-532
Supplier:  Genetex
Supplier Number:  GTX104763

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD274 molecule
  • Antigen Symbol:
    CD274
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    29126
  • Antigen Synonyms:
    PDL1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
  • Storage Buffer:
    1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.
  • Molecular Weight:
    33 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    3.47mg/ml
  • Immunogen:
    Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PD-L1. The exact sequence is proprietary.
  • Tested Applications:
    IHC-P
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89350-532
  • Supplier no.:
    GTX104763

Specifications

About this item

Rabbit Polyclonal antibody to PD-L1 (CD274 molecule)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD274
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human