Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]
Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]
Catalog # 89353-374
Supplier:  Genetex
Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]
Catalog # 89353-374
Supplier:  Genetex
Supplier Number:  GTX75791

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Beta 2
  • Antigen Symbol:
    ITGB2
  • Clonality:
    Monoclonal
  • Clone:
    MEM-148
  • Conjugation:
    PE (Phycoerythrin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 test
  • Gene ID:
    3689
  • Antigen Synonyms:
    LFA-1|CD18|MAC-1|integrin subunit beta 2|LAD|LCAMB|MFI7|MF17
  • Storage Buffer:
    Phosphate buffered saline containing 0.2% bovine serum albumin and 0.09% sodium azide
  • Storage Temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
  • Immunogen:
    Peripheral blood mononuclear cells
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Cat. No.:
    89353-374
  • Supplier no.:
    GTX75791

Specifications

About this item

Mouse Antibody to Human CD18(ACT Epitope):RPE

Flow Cytometry: Use 20ul of the suggested working dilution to label 10^6 cells in 100ul. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human