Anti-CD172A Mouse Monoclonal Antibody [clone: ED9]
89367-172
: Genetex
- Antibody Type:Primary
- Antigen Name:Signal-regulatory Protein Alpha
- Antigen Symbol:SIRPA
- Clonality:Monoclonal
- Clone:ED9
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:50 test
- Gene ID:25528
- Antigen Synonyms:Signal-regulatory protein alpha|PTPNS1|CD172A|MYD-1|SHPS1|signal regulatory protein alpha|P84|BIT|SIRP|MFR
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
- Storage Temperature:Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Spleen cell homogenate
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. No.:89367-172
- Supplier no.:GTX76362
Mouse monoclonal antibody [ED9] to SIRP alpha
Type: Primary
Antigen: CD172A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: