Anti-BAK Mouse Monoclonal Antibody [clone: TC100]
89355-276
: Genetex
- Antibody Type:Primary
- Antigen Name:BCL2-Antagonist/Killer 1
- Antigen Symbol:BAK
- Clonality:Monoclonal
- Clone:TC100
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Gene ID:578
- Antigen Synonyms:bcl-2 homologous antagonist/killer|bcl-2-like protein 7|BCL2-like 7 protein|pro-apoptotic protein BAK|BAK1|bcl2-L-7|apoptosis regulator BAK
- Storage Buffer:Preservative: 0.09% Sodium Azide; Constituents: 0.2% Gelatin, 50mM Sodium Phosphate
- Storage Temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:0.1mg/ml
- Immunogen:Recombinant fragment (C-terminal transmembrane portion deleted) (Human).
- Tested Applications:IHC-Fr, WB
- Purification:Immunogen affinity purified
- Cat. No.:89355-276
- Supplier no.:GTX10808
Mouse monoclonal [TC100] to Bak
IHC-Fr: Use at a concentration of 1 - 5 ?g/ml. WB: Use at a concentration of 2 - 5 ?g/ml. Detects a band of approximately 30 kDa (predicted molecular weight: 23.4 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: BAK
Clonality: Monoclonal
Clone: TC100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human