Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AMPD2 Rabbit Polyclonal Antibody
Anti-AMPD2 Rabbit Polyclonal Antibody
Catalog # 89357-086
Supplier:  Genetex
Anti-AMPD2 Rabbit Polyclonal Antibody
Catalog # 89357-086
Supplier:  Genetex
Supplier Number:  GTX112483

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AMP deaminase 2
  • Antigen Symbol:
    AMPD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    271
  • Antigen Synonyms:
    PCH9|SPG63
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Molecular Weight:
    101 kDa
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
  • Concentration:
    0.99mg/ml
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human AMPD2. The exact sequence is proprietary.
  • Tested Applications:
    IHC-P
  • Purification:
    Purified by antigen-affinity chromatography.
  • Cat. No.:
    89357-086
  • Supplier no.:
    GTX112483

Specifications

About this item

Rabbit Polyclonal antibody to AMPD2 (adenosine monophosphate deaminase 2)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AMPD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human