Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AKR1B10 Mouse Monoclonal Antibody [clone: B981M]
Anti-AKR1B10 Mouse Monoclonal Antibody [clone: B981M]
Catalog # 89363-538
Supplier:  Genetex
Anti-AKR1B10 Mouse Monoclonal Antibody [clone: B981M]
Catalog # 89363-538
Supplier:  Genetex
Supplier Number:  GTX44456

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Aldo-keto reductase family 1 member B10
  • Antigen Symbol:
    AKR1B10
  • Clonality:
    Monoclonal
  • Clone:
    B981M
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    200 μg
  • Gene ID:
    57016
  • Antigen Synonyms:
    aldo-keto reductase family 1|ALDRLn|AKR1B11|ARL-1|small intestine reductase|AKR1B12|aldose reductase-like 1N: aldose reductase-like peptide|aldose reductase-related protein|AKR1B10|ARL1|member B10 (aldose reductase) |member B10|MGC14103|HIS|member B11 (aldose reductase-like)
  • Storage Temperature:
    Upon receipt, store at –20°C. Avoid multiple freeze/thaw cycles.
  • Tested Applications:
    ELISA
  • Cat. No.:
    89363-538
  • Supplier no.:
    GTX44456

Specifications

About this item

Mouse Monoclonal antibody [B981M] to AKR1B10

Type: Primary
Antigen: AKR1B10
Clonality: Monoclonal
Clone: B981M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human