Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Catalog # 89351-548
Supplier:  Genetex
Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Catalog # 89351-548
Supplier:  Genetex
Supplier Number:  GTX20785

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Melan A
  • Antigen Symbol:
    MLANA
  • Clonality:
    Monoclonal
  • Clone:
    A103
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    500 μl
  • Gene ID:
    2315
  • Antigen Synonyms:
    MART-1|MART1
  • Storage Buffer:
    Phosphate-buffered saline, pH 7.2, containing 0.09% sodium azide
  • Storage Temperature:
    Keep as concentrated solution, aliquot and store at 4°C.
  • Immunogen:
    Recombinant full length protein (Human).
  • Tested Applications:
    IHC-P, IP, WB
  • Purification:
    Purified
  • Cat. No.:
    89351-548
  • Supplier no.:
    GTX20785

Specifications

About this item

MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes.

For IHC-P: Use at a dilution of 1:50-1:100 in an ABC method. We suggest an incubation period of 30 minutes at RT. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. For IP: Use at a dilution of 1:50. For WB: Use at a dilution of 1:50-1:100. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: A103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse