Anti-IgG Rabbit Polyclonal Antibody
89352-122
: Genetex
- Antibody Type:Primary
- Antigen Name:Immunoglobulin G
- Antigen Symbol:IgG
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:500 μl
- Storage Buffer:PBS with sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
- Immunogen:IgG isolated from human serum.
- Tested Applications:ICC/IF
- Purification:IgG fraction
- Cat. No.:89352-122
- Supplier no.:GTX24220
Rabbit polyclonal to Human IgG (FITC)
Immunofluorescence: use at a dilution of 1:20-1:40 for 30 - 60 mins at RT. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human