To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Carbohydrate-responsive element-binding protein
- Antigen Symbol:ChREBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:51085
- Antigen Synonyms:WBSCR14|WS-bHLH|MIO|MONDOB|Williams Beuren Syndrome Chromosome Region 14
- Storage Buffer:Contains 0.1% sodium azide
- Storage Temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Concentration:1.2 mg/ml
- Immunogen:A C-terminal synthetic peptide made to the human ChREBP protein sequence (between residues 800-852). NP_116569.1, isoform 1/alpha
- Tested Applications:ICC/IF, IHC-P, WB
- Purification:Affinity purified with antigen
- Cat. No.:89367-098
- Supplier no.:GTX30677
Rabbit polyclonal antibody to ChREBP
Recommended Starting Dilutions:For IF: Use at a dilution of 1:100-1:500. For IHC-P: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: ChREBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: