Anti-IL4 Rabbit Polyclonal Antibody
Catalog # 89365-374
Supplier: Genetex
Specifications
- Antibody Type:Primary
- Antigen Name:Interleukin 4
- Antigen Symbol:IL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:16189
- Antigen Synonyms:BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
- Storage Buffer:Phosphate buffered saline
- Storage Temperature:Prior to reconstitution store at +4 °C. Following reconstitution store at –20 °C. Store undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Recombinant mouse IL4
- Tested Applications:Functional Assay
- Purification:Purified IgG was prepared by affinity chromatography.
- Cat. No.:89365-374
- Supplier no.:GTX74063
Specifications
About this item
Rabbit polyclonal antibody to IL4
Recommended Starting Dilutions: ELISA: Max Dilution 0.5 ug/ml Functional Assays: 0.25 ug/ml - 0.40 ug/ml Western Blotting: 0.1 ug/ml - 0.2 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by
Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse