Quick Disconnect Connector, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Devices enable quick disconnection of high-density 3–12 mm I.D
polyethylene tubing. Tubing slides snugly together quickly and easily. An important safety factor on vacuum lines used with filtering flasks.