Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Titration-Child-Page22.gif
VeriSpec® Certified Reference Materials
RICCA Chemical Company provides standards for AA, ICP, ICP/MS, Ion Chromatography, pH Calibration Buffers, Conductivity and PFAS analysis that meet ISO 17034 and ISO 17025 requirements with our premium line of VeriSpec® Certified Reference Materials. These products have undergone rigorous evaluations of manufacturing methods, quality control, stability, and homogeneity. As a result, a certificate with the specified property value, uncertainty, and traceability is available with every bottle.
Titrants | Avantor
Source with RICCA and you can expect:
  • The highest level of quality assurance
  • Increased confidence in your analyses
  • Technical support every step of the way
  • Detailed C of A’s that provide traceability of measurements & calibration to national standards
  • Full Line of Single, Multi-Element, and Multi-Ion Solutions
  • EPA Contract Laboratory Program (CLP)
  • Custom blends to meet your requirements
  • 50 years successfully serving scientists
Product selection
EDTA1.png
Ethylenediaminetetraacetic acid, the all-purpose chelating agent perfect for metals determination.
Sodium-Hydroxide2.png
NaOH & KOH options from 0.02N – 1.00 N, tested in ISO 17025 accredited laboratories.
VeriSpec Bottles 2.png
Unleash controlled reactivity for precise titrations, ensuring accuracy and excellence in every chemical analysis with self-indicating KMnO4.
PFAS vial and box.png
Ensure accuracy and regulatory compliance in your PFAS testing with our CRMs