Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
honeywell_oligonucleotide_synthesis_hero.jpg
Solvents and Reagents for Oligonucleotide Synthesis
Burdick & Jackson™ BioSyn™ solvents and reagents for DNA and RNA synthesis are processed and purified to minimize particulate contamination, making them ideal for the high purity required in pharmacogenomics, diagnostics, therapeutics and drug discovery applications.
Solvents and Reagents for Oligonucleotide Synthesis​ | Avantor
These solvents and reagents are ideal for use in performing oligosynthesis at lab scale or production levels and are available in a range of package sizes from 1L glass bottles to 1,250L bulk containers. Packaging is designed to facilitate direct connection to popular synthesizers.
Product selection
400x495_Deblocking_reagents.jpg
Deblocking reagents for detritylation in various formulations.
400x495_Activatorreagents_1.jpg
High performance activator solutions for oligonucleotide synthesis.
400x495_Activatorreagents.jpg
Acetic anhydride or N-Methylimidazole solution for the capping step.
400x495_Oxidation_reagents.jpg
Reagents for the oxidation step in the oligonucleotide synthesis cycle.
400x495_Solvents_related_products.jpg
Solvents and related products for the oligonucleotide synthesis including solvents with low water content.