Trident LC Column Protection System
Catalog # REST27473
Supplier: Restek
New Product
Specifications
- Opis:Trident column protection system, with Level 2 - cartridge holder only
- Ilość:4
Specifications
About this item
Trident LC column protection system is available in three levels of protection such as filter only, cartridge only, or filter and cartridge.
- Easy-to-remove cap frit simplifies filter replacement
- Low-dead-volume design has negligible effect on chromatography
- Direct connection eliminates tubing and connectors that increase system volume and leak potential
The durable metal tip with replaceable PEEK ferrule means easy installation onto column without tools. Improved thread design and materials create an optimal seal that releases and reseals easily, allowing multiple installations without galling and binding.