Real Time RT-PCR Kit
Catalog # GSDCPCOV6111
Supplier: GOLD STANDARD DIAGNOSTICS
CAS Number:
Specifications
- Ilość:1 KIT
- Opis:GSD NovaPrime® TSP SARS-CoV-2, 1×96 reactions
Specifications
About this item
High quality RT-PCR kits for detection of various infectious diseases.
- Fast and reliable results in approximately 1 hour
- Validated for a variety of specimen types
- Excellent performance with high sensitivity and specificity
- Extraction/inhibition and positive control included
- Suitable for most PCR platforms